Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acox57D-d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079079.4 | NP_523803.1 | 1 - 2 | 236 | 68 | |
GenBank mRNA | AY070539.1 | - | 1 - 2 | 245 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |