Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fd59A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079090.3 | NP_523814.1 | 1 - 2 | 696 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |