Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: key | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259574.2 | NP_001246503.1 | 3 - 4 | 586 | 66 | |
NM_079132.4 | NP_523856.2 | 3 - 4 | 586 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |