Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NaCP60E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202078.3 | NP_001189007.2 | 11 - 12 | 3253 | 87 | |
NM_001259578.1 | NP_001246507.1 | 8 - 9 | 2354 | 87 | ||
NM_001259579.3 | NP_001246508.1 | 11 - 12 | 3516 | 87 | ||
NM_001259580.2 | NP_001246509.1 | 11 - 12 | 3253 | 87 | ||
NM_001259581.2 | NP_001246510.1 | 11 - 12 | 3516 | 87 | ||
NM_001274243.2 | NP_001261172.1 | 11 - 12 | 3253 | 87 | ||
NM_001274244.2 | NP_001261173.1 | 11 - 12 | 3253 | 87 | ||
NM_001274245.2 | NP_001261174.1 | 10 - 11 | 3310 | 87 | ||
NM_001274246.2 | NP_001261175.1 | 11 - 12 | 3253 | 87 | ||
NM_166696.5 | NP_726495.3 | 11 - 12 | 3516 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |