Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043111.3 | NP_001036576.1 | 6 - 7 | 2431 | 78 | |
NM_001103993.2 | NP_001097463.1 | 6 - 7 | 2431 | 78 | ||
NM_001274282.2 | NP_001261211.1 | 5 - 6 | 2337 | 78 | ||
NM_001274283.2 | NP_001261212.1 | 2 - 3 | 1253 | 78 | ||
NM_001299959.1 | NP_001286888.1 | 2 - 3 | 1253 | 78 | ||
NM_079149.5 | NP_523873.2 | 6 - 7 | 2431 | 78 | ||
GenBank mRNA | BT010042.1 | - | 6 - 7 | 2021 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |