Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259801.2 | NP_001246730.1 | 2 - 3 | 1183 | 97 | |
NM_079309.4 | NP_524033.2 | 2 - 3 | 1183 | 97 | ||
GenBank mRNA | AY061440.1 | - | 2 - 3 | 1189 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |