Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169984.3 | NP_001163455.2 | 3 - 4 | 1330 | 81 | |
NM_001259871.1 | NP_001246800.1 | 3 - 4 | 1478 | 81 | ||
NM_001259872.1 | NP_001246801.1 | 3 - 4 | 1478 | 81 | ||
NM_001259873.1 | NP_001246802.1 | 3 - 4 | 1330 | 81 | ||
NM_001275038.1 | NP_001261967.1 | 3 - 4 | 1478 | 81 | ||
NM_001275039.1 | NP_001261968.1 | 3 - 4 | 1330 | 81 | ||
NM_001275040.1 | NP_001261969.1 | 3 - 4 | 1330 | 81 | ||
NM_079395.4 | NP_524119.2 | 3 - 4 | 1330 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |