Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275142.1 | NP_001262071.1 | 4 - 5 | 1503 | 78 | |
NM_079446.4 | NP_524170.2 | 4 - 5 | 1491 | 78 | ||
GenBank mRNA | AY051880.1 | - | 4 - 5 | 1460 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |