Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144525.2 | NP_001137997.2 | 7 - 8 | 1562 | 70 | |
NM_079485.4 | NP_524209.3 | 7 - 8 | 1565 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |