Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ih | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038856.4 | NP_001033945.1 | 4 - 5 | 4579 | 68 | |
NM_001038858.4 | NP_001033947.1 | 2 - 3 | 5291 | 68 | ||
NM_001038859.4 | NP_001033948.1 | 3 - 4 | 4537 | 68 | ||
NM_001144195.3 | NP_001137667.1 | 3 - 4 | 4537 | 68 | ||
NM_001144196.3 | NP_001137668.1 | 3 - 4 | 4537 | 68 | ||
NM_001144197.3 | NP_001137669.1 | 3 - 4 | 4537 | 68 | ||
GenBank mRNA | AY060342.1 | - | 1 - 2 | 1309 | 68 | |
AY928070.1 | - | 4 - 5 | 2040 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |