Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oaz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103845.3 | NP_001097315.1 | 1 - 2 | 341 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |