Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spred | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274049.2 | NP_001260978.1 | 3 - 4 | 622 | 112 | |
NM_001274050.2 | NP_001260979.1 | 3 - 4 | 688 | 112 | ||
NM_137144.4 | NP_610988.1 | 3 - 4 | 688 | 112 | ||
GenBank mRNA | BT126047.1 | - | 2 - 3 | 287 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |