Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir52a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137197.2 | NP_611041.2 | 1 | 1760 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |