Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258601.3 | NP_001245530.1 | 2 - 3 | 327 | 92 | |
NM_001370042.1 | NP_001356933.1 | 2 - 3 | 327 | 92 | ||
NM_080336.5 | NP_525075.1 | 2 - 3 | 327 | 92 | ||
NM_167014.3 | NP_726942.1 | 2 - 3 | 441 | 92 | ||
NM_167015.3 | NP_726943.1 | 2 - 3 | 682 | 92 | ||
GenBank mRNA | KX531290.1 | - | 1 - 2 | 112 | 92 | |
U32855.1 | - | 2 - 3 | 682 | 92 | ||
U48846.1 | - | 2 - 3 | 309 | 92 | ||
U48848.1 | - | 2 - 3 | 313 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |