Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298032.1 | NP_001284961.1 | 1 - 2 | 112 | 102 | |
NM_132118.3 | NP_572346.1 | 2 - 3 | 382 | 102 | ||
NM_167087.1 | NP_727118.1 | 2 - 3 | 119 | 102 | ||
NM_167088.2 | NP_727119.1 | 2 - 3 | 136 | 102 | ||
NM_167089.2 | NP_727120.1 | 2 - 3 | 223 | 102 | ||
GenBank mRNA | AY060845.1 | - | 2 - 3 | 347 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |