Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4g15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132493.3 | NP_572721.1 | 4 - 5 | 1806 | 86 | |
NM_167287.2 | NP_727531.2 | 4 - 5 | 1806 | 86 | ||
GenBank mRNA | AY060719.1 | - | 4 - 5 | 1783 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |