Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Optix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273864.2 | NP_001260793.1 | 1 - 2 | 960 | 127 | |
NM_079956.3 | NP_524695.2 | 1 - 2 | 650 | 127 | ||
GenBank mRNA | AY061077.1 | - | 1 - 2 | 610 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |