Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ect | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300090.1 | NP_001287019.1 | 4 - 5 | 808 | 110 | |
NM_079967.6 | NP_524706.2 | 4 - 5 | 841 | 110 | ||
NM_168400.3 | NP_729596.1 | 4 - 5 | 784 | 110 | ||
NM_168401.3 | NP_729597.2 | 4 - 5 | 841 | 110 | ||
GenBank mRNA | AY061556.1 | - | 4 - 5 | 825 | 110 | |
BT003781.1 | - | 4 - 5 | 837 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |