Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Roc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144671.3 | NP_001138143.2 | 1 - 2 | 332 | 54 | |
NM_001297825.1 | NP_001284754.1 | 2 - 3 | 248 | 54 | ||
NM_130496.4 | NP_569852.1 | 2 - 3 | 248 | 54 | ||
GenBank mRNA | AY119265.1 | - | 2 - 3 | 265 | 54 | |
KX531597.1 | - | 2 - 3 | 104 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |