Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp53C14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299572.1 | NP_001286501.1 | 1 - 2 | 84 | 68 | |
NM_137308.3 | NP_611152.1 | 1 - 2 | 84 | 68 | ||
GenBank mRNA | KX532084.1 | - | 1 - 2 | 41 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |