Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp53C14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299573.1 | NP_001286502.1 | 1 - 2 | 81 | 101 | |
NM_137309.4 | NP_611153.1 | 1 - 2 | 81 | 101 | ||
GenBank mRNA | BT023638.1 | - | 1 - 2 | 63 | 101 | |
BT032660.1 | - | 1 - 2 | 68 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |