Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274120.2 | NP_001261049.1 | 2 - 3 | 919 | 60 | |
NM_001299596.1 | NP_001286525.1 | 2 - 3 | 1077 | 60 | ||
NM_137364.4 | NP_611208.1 | 2 - 3 | 919 | 60 | ||
GenBank mRNA | BT004912.1 | - | 2 - 3 | 894 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |