Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGAP54D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274131.2 | NP_001261060.1 | 5 - 6 | 2682 | 92 | |
NM_137392.3 | NP_611236.2 | 5 - 6 | 2878 | 92 | ||
GenBank mRNA | BT133065.1 | - | 5 - 6 | 2666 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |