Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jheh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137541.4 | NP_611385.1 | 3 - 4 | 653 | 83 | |
GenBank mRNA | AF517545.1 | - | 3 - 4 | 619 | 83 | |
BT001753.1 | - | 3 - 4 | 676 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |