Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272562.1 | NP_001259491.1 | 9 - 10 | 9444 | 58 | |
NM_080536.4 | NP_536797.3 | 8 - 9 | 8825 | 58 | ||
NM_167334.2 | NP_727634.2 | 8 - 9 | 8739 | 58 | ||
NM_167335.2 | NP_727635.2 | 9 - 10 | 9504 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |