Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298310.1 | NP_001285239.1 | 7 - 8 | 1471 | 61 | |
NM_167408.2 | NP_727781.1 | 8 - 9 | 1592 | 61 | ||
GenBank mRNA | AY060637.1 | - | 8 - 9 | 1592 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |