Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pyx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138171.2 | NP_612015.1 | 7 - 8 | 2358 | 92 | |
NM_167813.2 | NP_728501.1 | 3 - 4 | 894 | 92 | ||
GenBank mRNA | BT001363.1 | - | 3 - 4 | 894 | 92 | |
BT010317.1 | - | 7 - 8 | 2358 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster Inventoried |