Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272173.1 | NP_001259102.1 | 3 - 4 | 442 | 100 | |
NM_130502.3 | NP_569858.1 | 3 - 4 | 442 | 100 | ||
GenBank mRNA | AB176551.1 | - | 3 - 4 | 283 | 100 | |
AY095525.1 | - | 3 - 4 | 397 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |