Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137828.1 | NP_611672.1 | 1 - 2 | 147 | 69 | |
GenBank mRNA | AY226543.1 | - | 1 - 2 | 147 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |