Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir60e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138083.3 | NP_611927.3 | 1 | 1604 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |