Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138087.6 | NP_611931.3 | 3 - 4 | 2719 | 89 | |
GenBank mRNA | AY060468.1 | - | 3 - 4 | 2684 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |