Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298007.1 | NP_001284936.1 | 2 - 3 | 237 | 78 | |
NM_143705.3 | NP_651962.2 | 2 - 3 | 237 | 78 | ||
GenBank mRNA | BT150092.1 | - | 2 - 3 | 211 | 78 | |
BT150473.1 | - | 2 - 3 | 211 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |