Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ScsbetaG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300012.1 | NP_001286941.1 | 3 - 4 | 428 | 59 | |
NM_143706.5 | NP_651963.1 | 3 - 4 | 428 | 59 | ||
GenBank mRNA | AJ252069.1 | - | 3 - 4 | 395 | 59 | |
AY051980.1 | - | 3 - 4 | 380 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |