Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpmeg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169838.3 | NP_001163309.2 | 6 - 7 | 1808 | 60 | |
NM_001169839.3 | NP_001163310.2 | 6 - 7 | 1929 | 60 | ||
NM_138187.4 | NP_612031.3 | 6 - 7 | 1820 | 60 | ||
NM_167830.4 | NP_728522.2 | 6 - 7 | 1980 | 60 | ||
NM_167831.3 | NP_728523.2 | 7 - 8 | 2017 | 60 | ||
GenBank mRNA | BT044533.1 | - | 6 - 7 | 1888 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |