Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHGPx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168024.2 | NP_728868.1 | 1 - 2 | 445 | 68 | |
GenBank mRNA | BT011331.1 | - | 1 - 2 | 177 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |