Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ero1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139608.4 | NP_647865.2 | 1 - 2 | 388 | 67 | |
GenBank mRNA | BT021422.1 | - | 1 - 2 | 336 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |