Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298034.1 | NP_001284963.1 | 7 - 8 | 1601 | 71 | |
NM_001298035.1 | NP_001284964.1 | 7 - 8 | 1538 | 71 | ||
NM_001298036.1 | NP_001284965.1 | 7 - 8 | 1550 | 71 | ||
NM_132121.3 | NP_572349.1 | 7 - 8 | 1589 | 71 | ||
GenBank mRNA | AY051622.1 | - | 7 - 8 | 1589 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |