Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144438.2 | NP_001137910.2 | 3 | 1872 | 109 | |
NM_001144439.2 | NP_001137911.1 | 3 | 1797 | 109 | ||
NM_143787.3 | NP_652044.1 | 3 | 1842 | 109 | ||
NM_168265.2 | NP_729366.1 | 2 | 1924 | 109 | ||
NM_168266.2 | NP_729367.1 | 2 | 1540 | 109 | ||
NM_168267.2 | NP_729368.1 | 2 | 1514 | 109 | ||
NM_168268.3 | NP_729369.2 | 3 | 1674 | 109 | ||
NM_168269.2 | NP_729370.1 | 2 | 1544 | 109 | ||
NM_176298.2 | NP_788476.1 | 3 | 1707 | 109 | ||
GenBank mRNA | AY047505.1 | - | 2 | 1931 | 109 | |
BT099506.1 | - | 2 | 1517 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |