Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169916.2 | NP_001163387.2 | 17 - 18 | 7623 | 84 | |
NM_001202142.2 | NP_001189071.1 | 15 - 16 | 4206 | 84 | ||
NM_001202143.2 | NP_001189072.1 | 15 - 16 | 4206 | 84 | ||
NM_001259753.2 | NP_001246682.1 | 14 - 15 | 3885 | 84 | ||
NM_001259754.2 | NP_001246683.1 | 14 - 15 | 3483 | 84 | ||
NM_001259755.1 | NP_001246684.1 | 15 - 16 | 5208 | 84 | ||
NM_001274670.1 | NP_001261599.1 | 17 - 18 | 8147 | 84 | ||
NM_168293.3 | NP_729410.1 | 15 - 16 | 4206 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |