Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104145.3 | NP_001097615.1 | 4 - 5 | 1407 | 68 | |
NM_001259856.2 | NP_001246785.1 | 4 - 5 | 1688 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |