Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fend | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272443.2 | NP_001259372.1 | 3 - 4 | 1448 | 75 | |
NM_132300.3 | NP_572528.1 | 3 - 4 | 1389 | 75 | ||
GenBank mRNA | AB065095.1 | - | 3 - 4 | 1368 | 75 | |
AF209858.1 | - | 3 - 4 | 1367 | 75 | ||
BT030985.1 | - | 3 - 4 | 945 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |