Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4g15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132493.3 | NP_572721.1 | 1 - 2 | 732 | 71 | |
NM_167287.2 | NP_727531.2 | 1 - 2 | 732 | 71 | ||
GenBank mRNA | AY060719.1 | - | 1 - 2 | 709 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |