Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103493.4 | NP_001096963.1 | 3 - 4 | 3622 | 80 | |
NM_001103494.2 | NP_001096964.1 | 3 - 4 | 3741 | 80 | ||
NM_001298228.1 | NP_001285157.1 | 3 - 4 | 3741 | 80 | ||
NM_001298229.1 | NP_001285158.1 | 4 - 5 | 4056 | 80 | ||
NM_001298230.1 | NP_001285159.1 | 4 - 5 | 4056 | 80 | ||
NM_132584.3 | NP_572812.3 | 4 - 5 | 3930 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |