Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ArgRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132853.3 | NP_573081.1 | 3 - 4 | 572 | 83 | |
GenBank mRNA | AY070924.1 | - | 3 - 4 | 563 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |