Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272656.2 | NP_001259585.1 | 8 - 9 | 4942 | 65 | |
NM_001272657.2 | NP_001259586.1 | 8 - 9 | 4862 | 65 | ||
NM_001298376.1 | NP_001285305.1 | 8 - 9 | 4964 | 65 | ||
NM_132858.3 | NP_573086.2 | 8 - 9 | 4967 | 65 | ||
GenBank mRNA | AY118524.1 | - | 8 - 9 | 4942 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |