Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Teh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139609.2 | NP_647866.2 | 4 - 5 | 1277 | 87 | |
GenBank mRNA | BT022560.1 | - | 4 - 5 | 674 | 87 | |
DQ139960.1 | - | 4 - 5 | 650 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |