Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104031.2 | NP_001097501.1 | 9 - 10 | 1784 | 85 | |
NM_139636.4 | NP_647893.2 | 9 - 10 | 1672 | 85 | ||
GenBank mRNA | BT050431.1 | - | 9 - 10 | 1690 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |