Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176284.3 | NP_788462.2 | 4 - 5 | 1203 | 64 | |
GenBank mRNA | BT044127.1 | - | 4 - 5 | 1204 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Drosophila melanogaster |