Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139924.3 | NP_648181.2 | 1 - 2 | 665 | 115 | |
NM_176297.1 | NP_788475.1 | 2 - 3 | 617 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Drosophila melanogaster |