Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139947.3 | NP_648204.1 | 2 - 3 | 749 | 74 | |
GenBank mRNA | AY070491.1 | - | 2 - 3 | 746 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |